Learn More
TARGETMOL CHEMICALS INC BISDEMETHOXYCURCUMIN 50MG
SDP
Catalog No. 502576181
Encompass_Preferred
Also available in 5 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Bisdemethoxycurcumin (Curcumin III) is a natural demethoxy derivative of curcumin. It is a potent activator of macrophage phagocytosis interacting with 1(alpha) 25-dihydroxy vitamin D3 to stimulate amyloid-(beta) clearance by macrophages (optimal stimulation at 100 nM BDMC) [1]. More stable than curcumin in physiological media BDMC suppresses proliferation of cancer cells [2]. It down-regulates the transcriptional coactivator p300 suppressing the Wnt/(beta)-catenin pathway and inhibits LPS induction of iNOS expression [3]. purity: 99%
Catalog No. 50-257-6181 Supplier TARGETMOL CHEMICALS INC Supplier No. TL000750MG
Also available in 5 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Bisdemethoxycurcumin (Curcumin III) is a natural demethoxy derivative of curcumin. It is a potent activator of macrophage phagocytosis interacting with 1(alpha) 25-dihydroxy vitamin D3 to stimulate amyloid-(beta) clearance by macrophages (optimal stimulation at 100 nM BDMC) [1]. More stable than curcumin in physiological media BDMC suppresses proliferation of cancer cells [2]. It down-regulates the transcriptional coactivator p300 suppressing the Wnt/(beta)-catenin pathway and inhibits LPS induction of iNOS expression [3]. purity: 99%