Learn More
Abcam Bisdemetho x ycurcumin, Apoptotic curcuminoid, 5MG
Catalog No. 502707071
Encompass_Preferred
MW 308.33 g/mol, Purity >98%. Apoptotic anticancer, anti-inflammatory and antioxidant curcuminoid. Inhibits wnt activity through activation of WIF-1. Induces demethylation through DNA methyltransferase 1 inhibition. Potent and competitive aldo-keto reductase inhibitor (Ki = 22 nM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7071 Supplier Abcam Supplier No. AB1209765MG
MW 308.33 g/mol, Purity >98%. Apoptotic anticancer, anti-inflammatory and antioxidant curcuminoid. Inhibits wnt activity through activation of WIF-1. Induces demethylation through DNA methyltransferase 1 inhibition. Potent and competitive aldo-keto reductase inhibitor (Ki = 22 nM).
The product is subject to the following: Abcam Restricted Use Statement