Learn More
Fine Science Tools BISHOP-HARMON FORCEP 1X2 TEETH
SDP
Catalog No. NC0575711
Encompass
Tip Shape: Straight/ Tips: 1x2 Teeth/ Tip Dimensions: 0.5 x 0.5mm/ Alloy / Material: Stainless Steel/ Length: 8.5cm
Catalog No. NC0575711 Supplier Fine Science Tools Supplier No. 1107008
Tip Shape: Straight/ Tips: 1x2 Teeth/ Tip Dimensions: 0.5 x 0.5mm/ Alloy / Material: Stainless Steel/ Length: 8.5cm