Learn More
Stoelting™ Bishop-Harmon Forcep
SDP
Catalog No. 10001199 Shop All Stoelting Co Products
Change view
Click to view available options
Size:
3.25 in., 1 x 2.3
3.25 in., Cross Serrated; 0.5 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
10-001-199 3.25 in., Cross Serrated; 0.5 mm
10-001-200 3.25 in., 1 x 2.3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-001-199 Supplier Stoelting™ Supplier No. 5210275
Only null left
Add to Cart
Add to Cart
Micro tweezers are useful in a variety of different applications. Common uses include micro dissection of tissue and vessels, and processing tissue in electron microscopy procedures.
Specifications
Size 3.25 in., Cross Serrated; 0.5 mm
Model Fine