Learn More
World Precision Instrument Bishop-Harmon Forceps, 9cm, 1x2 Teeth, Titanium
SDP
Supplier:  World Precision Instrument WP3510
Encompass
Bishop Harmon Iris Forceps are a pair of surgical forceps (tweezers) often used in ophthalmic procedures. They are designed specifically for use in the anterior portion of the eye to reach structures like the iris. Iris forceps are a small pair of forceps (9cm long) with a fine tip for grasping delicate tissue with minimal trauma. You can choose between serrated tips and tips with a 1x2 tooth configuration. The narrow, serrated jaws of the surgical forceps allow for fine manipulation of structures like the iris of the eye. The 1x2 teeth give a secure grip of delicate structures with minimal trauma. Researchers like these fine tipped tweezers for precision work in microdissection. These forceps are have straight tips. The Iris Forceps are manufactured with surgical grade stainless steel, premium German steel, or titanium, all of which are autoclavable. WPI recommends properly cleaning all your surgical instruments promptly after use.
Catalog No. 50-253-8316