Learn More
Sklar Instruments Forceps, Sklar Bishop-Harmon Ophthalmic Tissue Forceps, Sklar 3-1/2 in., 1x2 Teeth, Straight, Smooth Jaws, Stainless Steel, EA.
SDP
Catalog No. 501118807 Shop All Sklar Instruments Products
Encompass_Preferred
Forceps, Sklar Bishop-Harmon Ophthalmic Tissue Forceps, Sklar, 3-1/2 in., 1x2 Teeth, Straight, Smooth Jaws, Stainless Steel, EA. Sklar Bishop-Harmon Ophthalmic Tissue Forceps are primarily used in ophthalmic procedures or microsurgery. They are delicate forceps with 1x2 fine teeth. Standard Bishop-Harmon forceps have fenestrated handles. This product is straight with 1x2 teeth and a length of 3-1/2 inches. Lifetime warranty against all manufacturer defects.
Catalog No. 50-111-8807 Supplier Sklar Instruments Supplier No. 664132
Forceps, Sklar Bishop-Harmon Ophthalmic Tissue Forceps, Sklar, 3-1/2 in., 1x2 Teeth, Straight, Smooth Jaws, Stainless Steel, EA. Sklar Bishop-Harmon Ophthalmic Tissue Forceps are primarily used in ophthalmic procedures or microsurgery. They are delicate forceps with 1x2 fine teeth. Standard Bishop-Harmon forceps have fenestrated handles. This product is straight with 1x2 teeth and a length of 3-1/2 inches. Lifetime warranty against all manufacturer defects.