Learn More
Enzo Life Sciences Bisidolylmaleimide I hydrochloride (1mg). CAS: 176504-36-2
SDP
Catalog No. 502013381
Encompass_Preferred
Enhanced, water soluble form of Bisindolylmaleimide I. Alternative name: 3-[1-(Dimethylaminopropyl)indol-3-yl]-4-(indol-3-yl)maleimide hydrochloride. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO or methanol. Long Term Storage: -80°C.
Catalog No. 50-201-3381 Supplier Enzo Life Sciences Supplier No. ENZCHM1080001
Enhanced, water soluble form of Bisindolylmaleimide I. Alternative name: 3-[1-(Dimethylaminopropyl)indol-3-yl]-4-(indol-3-yl)maleimide hydrochloride. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO or methanol. Long Term Storage: -80°C.