Learn More
Cayman Chemical Bisindolylmaleimide IX (mesylate), Purity: ≥98%, CAS: 138489-18-6, 10 mg
SDP
Catalog No. 502951869
Encompass_Preferred
A potent, cell-permeable inhibitor of PKC isoforms (IC50 = 5, 24, 14, 27, and 24 nM for PKC-α, PKC-βI, PKC-βII, PKC-γ, and PKC-ε, respectively)
Catalog No. 50-295-1869 Supplier Cayman Chemical Supplier No. 1333410 MG
A potent, cell-permeable inhibitor of PKC isoforms (IC50 = 5, 24, 14, 27, and 24 nM for PKC-α, PKC-βI, PKC-βII, PKC-γ, and PKC-ε, respectively)