Learn More
Cayman Chemical BIsIndolylmaleImID VIIIAC 10mg
SDP
Catalog No. 502952080
Encompass_Preferred
A selective PKC inhibitor (IC50 = 158 nM for rat brain PKC) that acts at the ATP binding site of PKC; exhibits PKC isozyme specificity with preference for PKCα over PKCβI, PKCβII, PKCγ, or PKCε (IC50s = 53, 195, 163, 213, and 175 nM, respectively)
Catalog No. 50-295-2080 Supplier Cayman Chemical Supplier No. 1333310 MG
A selective PKC inhibitor (IC50 = 158 nM for rat brain PKC) that acts at the ATP binding site of PKC; exhibits PKC isozyme specificity with preference for PKCα over PKCβI, PKCβII, PKCγ, or PKCε (IC50s = 53, 195, 163, 213, and 175 nM, respectively)