Learn More
Medchemexpress LLC Bisindolylmaleimide 100mg | 119139-23-0 | 100MG
SDP
Catalog No. 5000242780
Encompass_Preferred
Bisindolylmaleimide IV (Arcyriarubin A) is a potent protein kinase C (PKC) inhibitor with IC50s ranging from 0 1 to 0 55 M Bisindolylmaleimide IV also inhibits PKA (IC50 3 1-11 8 M)[1 Bisindolylmaleimide IV is a potent selective inhibitor of human cytomegalovirus (HCMV) replication in cell culture with an IC50 of 0 2 M[2
Catalog No. 50-002-42780 Supplier Medchemexpress LLC Supplier No. HY108254100MG
Bisindolylmaleimide IV (Arcyriarubin A) is a potent protein kinase C (PKC) inhibitor with IC50s ranging from 0 1 to 0 55 M Bisindolylmaleimide IV also inhibits PKA (IC50 3 1-11 8 M)[1 Bisindolylmaleimide IV is a potent selective inhibitor of human cytomegalovirus (HCMV) replication in cell culture with an IC50 of 0 2 M[2