Learn More
Medchemexpress LLC Bisindolylmaleimide I | 133052-90-1 | 412.48 | 25 MG
SDP
Catalog No. 5000348650
Encompass_Preferred
Bisindolylmaleimide I (GF109203X) is a cell-permeable and reversible protein kinase C (PKC) inhibitor. It exhibits IC50 values of 20 nM for PKCα, 17 nM for PKCβI, 16 nM for PKCβII, and 20 nM for PKCγ. This compound also functions as a glycogen synthase kinase-3 (GSK-3) inhibitor.
  • Cell-permeable and reversible protein kinase C inhibitor
  • Inhibits PKCα, PKCβI, PKCβII, and PKCγ with high potency
  • Functions as a GSK-3 inhibitor
  • Applicable in metabolic disease research
  • Applicable in cancer research
Catalog No. 50-003-48650 Supplier Medchemexpress LLC Supplier No. 25MGHY13867
Bisindolylmaleimide I (GF109203X) is a cell-permeable and reversible protein kinase C (PKC) inhibitor. It exhibits IC50 values of 20 nM for PKCα, 17 nM for PKCβI, 16 nM for PKCβII, and 20 nM for PKCγ. This compound also functions as a glycogen synthase kinase-3 (GSK-3) inhibitor.
  • Cell-permeable and reversible protein kinase C inhibitor
  • Inhibits PKCα, PKCβI, PKCβII, and PKCγ with high potency
  • Functions as a GSK-3 inhibitor
  • Applicable in metabolic disease research
  • Applicable in cancer research