Learn More
Selleck Chemical LLC Bisindolylmaleimide I
SDP
Catalog No. 5000004037
Encompass_Preferred
Bisindolylmaleimide I (GF109203X GO 6850) is a potent PKC inhibitor with IC50 of 20 nM 17 nM 16 nM and 20 nM for PKC PKC I PKC II and PKC in cell-free assays respectively showing more than 3000-fold selectivity for PKC as compared to EGFR PDGFR and insulin receptor
Catalog No. 50-000-04037 Supplier Selleck Chemical LLC Supplier No. S720810MM/1ML
Bisindolylmaleimide I (GF109203X GO 6850) is a potent PKC inhibitor with IC50 of 20 nM 17 nM 16 nM and 20 nM for PKC PKC I PKC II and PKC in cell-free assays respectively showing more than 3000-fold selectivity for PKC as compared to EGFR PDGFR and insulin receptor