Learn More
AdipoGen Bisindolylmaleimide I free b
SDP
Catalog No. 502562398
Encompass_Preferred
Chemical. CAS 133052-90-1. Formula C25H24N4O2. MW 412.5. Cell permeable kinase inhibitor with improved selectivity for protein kinase C PKC Ki = 10 nM. Competetive inhibitor for the ATP-binding site of PKC. Anti-inflammatory. Binds to P-glycoprotein. Telomerase activity inhibitor. Potent glycogen synthase kinase-3 GSK-3 inhibitor. Necrosis inhibitor. Blocks hERG potassium channels. Promotes osteoblastogenesis in human mesenchymal stem cells.
Catalog No. 50-256-2398 Supplier AdipoGen Supplier No. AGCR10009M001
Chemical. CAS 133052-90-1. Formula C25H24N4O2. MW 412.5. Cell permeable kinase inhibitor with improved selectivity for protein kinase C PKC Ki = 10 nM. Competetive inhibitor for the ATP-binding site of PKC. Anti-inflammatory. Binds to P-glycoprotein. Telomerase activity inhibitor. Potent glycogen synthase kinase-3 GSK-3 inhibitor. Necrosis inhibitor. Blocks hERG potassium channels. Promotes osteoblastogenesis in human mesenchymal stem cells.