Learn More
Medchemexpress LLC Bisindolylmaleimide I hydrochloride | 176504-36-2 | 98.4% | 448.94 | 50 MG
SDP
Catalog No. 5000349869
Encompass_Preferred
Bisindolylmaleimide I hydrochloride is a cell-permeable and reversible PKC inhibitor, with IC50 values of 20 nM for PKCα, 17 nM for PKCβI, 16 nM for PKCβII, and 20 nM for PKCγ. It also functions as a GSK-3 inhibitor. This product is intended for research use only.
  • Acts as a cell-permeable and reversible PKC inhibitor
  • Inhibits multiple PKC isoforms including alpha, beta I, beta II, and gamma
  • Demonstrates activity as a GSK-3 inhibitor
  • Capable of inhibiting alpha-thrombin-induced P47 phosphorylation
  • Inhibits DNA synthesis in quiescent cells
  • Reduces GSK-3 activity in adipocyte lysates
  • Inhibits exosome and microvesicle release from PC3 cells
  • Enhances the cytotoxicity of 5-fluorouracil
Catalog No. 50-003-49869 Supplier Medchemexpress LLC Supplier No. HY13867A50MG
Bisindolylmaleimide I hydrochloride is a cell-permeable and reversible PKC inhibitor, with IC50 values of 20 nM for PKCα, 17 nM for PKCβI, 16 nM for PKCβII, and 20 nM for PKCγ. It also functions as a GSK-3 inhibitor. This product is intended for research use only.
  • Acts as a cell-permeable and reversible PKC inhibitor
  • Inhibits multiple PKC isoforms including alpha, beta I, beta II, and gamma
  • Demonstrates activity as a GSK-3 inhibitor
  • Capable of inhibiting alpha-thrombin-induced P47 phosphorylation
  • Inhibits DNA synthesis in quiescent cells
  • Reduces GSK-3 activity in adipocyte lysates
  • Inhibits exosome and microvesicle release from PC3 cells
  • Enhances the cytotoxicity of 5-fluorouracil