Learn More
Medchemexpress LLC Bisindolylmaleimide I hydrochloride | 176504-36-2 | MFCD00236429 | 98.4% | 448.94 | C25H25ClN4O2 | 5 MG
SDP
Catalog No. 5000222156
Encompass_Preferred
Bisindolylmaleimide I hydrochloride is a cell-permeable, reversible protein kinase C (PKC) inhibitor that also shows activity against GSK-3. It is supplied as a powder for research and analytical use, with reported potency in the low nanomolar range against PKC isoforms and a high reported purity.
  • Cell-permeable, reversible PKC inhibitor
  • Potent activity: IC50 ≈ 16-20 nM for PKC isoforms
  • Also reported to inhibit GSK-3
  • High purity (reported 98.4%)
  • Supplied as a powder in small analytical pack sizes (for example, 1 mg, 5 mg, 10 mg)
  • Recommended storage: powder at 4°C or -20°C; solutions at -20°C or -80°C
Catalog No. 50-002-22156 Supplier Medchemexpress LLC Supplier No. HY13867A5MG
Bisindolylmaleimide I hydrochloride is a cell-permeable, reversible protein kinase C (PKC) inhibitor that also shows activity against GSK-3. It is supplied as a powder for research and analytical use, with reported potency in the low nanomolar range against PKC isoforms and a high reported purity.
  • Cell-permeable, reversible PKC inhibitor
  • Potent activity: IC50 ≈ 16-20 nM for PKC isoforms
  • Also reported to inhibit GSK-3
  • High purity (reported 98.4%)
  • Supplied as a powder in small analytical pack sizes (for example, 1 mg, 5 mg, 10 mg)
  • Recommended storage: powder at 4°C or -20°C; solutions at -20°C or -80°C