Learn More
Medchemexpress LLC Bisindolylmaleimide II | 137592-45-1 | MFCD00236430 | 99.8% | 438.52 | C27H26N4O2 | 5 MG
SDP
Catalog No. 5000264780
Encompass_Preferred
Bisindolylmaleimide II is a small-molecule inhibitor used to block protein kinase C (PKC) activity in biochemical and cellular research. Supplied as a solid with high stated purity and DMSO solubility, it is commonly used to probe PKC-dependent signaling and to evaluate kinase-related pathways in vitro.
  • General inhibitor of protein kinase C subtypes.
  • High purity for research applications.
  • Solid form facilitates accurate weighing and storage.
  • Soluble in DMSO for solution preparation.
  • Suitable for biochemical and cellular PKC assays.
Catalog No. 50-002-64780 Supplier Medchemexpress LLC Supplier No. HY1086045MG
Bisindolylmaleimide II is a small-molecule inhibitor used to block protein kinase C (PKC) activity in biochemical and cellular research. Supplied as a solid with high stated purity and DMSO solubility, it is commonly used to probe PKC-dependent signaling and to evaluate kinase-related pathways in vitro.
  • General inhibitor of protein kinase C subtypes.
  • High purity for research applications.
  • Solid form facilitates accurate weighing and storage.
  • Soluble in DMSO for solution preparation.
  • Suitable for biochemical and cellular PKC assays.