Learn More
AdipoGen Bisindolylmaleimide II (1 mg)
SDP
Catalog No. 502053758
Encompass_Preferred
Chemical. CAS: 137592-45-1. Formula: C27H26N4O2. MW: 438.5. Protein kinase C (PKC) inhibitor. Binds with reversed orientation to protein kinase A (PKA).
Catalog No. 50-205-3758 Supplier AdipoGen Supplier No. AGCR10010M001
Chemical. CAS: 137592-45-1. Formula: C27H26N4O2. MW: 438.5. Protein kinase C (PKC) inhibitor. Binds with reversed orientation to protein kinase A (PKA).