Learn More
Abcam Bisindolylmaleimide II, PKC inhibitor, 1MG
Catalog No. 502705993
Encompass_Preferred
MW 438.5 Da, Purity >97%. Potent, selective ATP-competitive PKC inhibitor (IC₅₀ values are 0.01, 0.75 and 2 μM for PKC, phosphorylase kinase and PKA respectively). Potent noncompetitive nAChR antagonist (IC₅₀ = 0.03 μM, catecholamine secretion in nicotine-stimulated PC-12 cells). Shows more potent antinicotinic effects than Bisindolylmaleimide IV (ab144208) and V (ab144205). Induces apoptosis and shows antiproliferative effects.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5993 Supplier Abcam Supplier No. AB1442061MG
MW 438.5 Da, Purity >97%. Potent, selective ATP-competitive PKC inhibitor (IC₅₀ values are 0.01, 0.75 and 2 μM for PKC, phosphorylase kinase and PKA respectively). Potent noncompetitive nAChR antagonist (IC₅₀ = 0.03 μM, catecholamine secretion in nicotine-stimulated PC-12 cells). Shows more potent antinicotinic effects than Bisindolylmaleimide IV (ab144208) and V (ab144205). Induces apoptosis and shows antiproliferative effects.
The product is subject to the following: Abcam Restricted Use Statement