Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bisindolylmaleimide II, Tocris Bioscience™
Description
Potent, ATP-competitive protein kinase C (PKC) inhibitor (IC50 = 0.01 μM). Displays selectivity for PKC over protein kinase A (PKA) and phosphorylase kinase (PK) (IC50 values are 0.75 and 2μM for PK and PKA respectively). Also displays potent, noncompetitive antagonism at nicotinic cholinergic receptors (IC50 ∽ 0.03 μM for inhibition of catecholamine secretion in nicotine-stimulated PC-12 cells).
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 438.52 |
| Percent Purity | >97% |
| Chemical Name or Material | Bisindolylmaleimide II |
Safety and Handling
| Recommended Storage | Store at -20°C |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?