Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bisindolylmaleimide II, Tocris Bioscience™
GSA_VA

Catalog No. p-4816577
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-281-0 10 mg
41-285-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 41-281-0 Supplier R&D Systems Supplier No. 4128/10
This item is not returnable. View return policy

Potent PKC inhibitor and nicotinic receptor antagonist

Potent, ATP-competitive protein kinase C (PKC) inhibitor (IC50 = 0.01 μM). Displays selectivity for PKC over protein kinase A (PKA) and phosphorylase kinase (PK) (IC50 values are 0.75 and 2μM for PK and PKA respectively). Also displays potent, noncompetitive antagonism at nicotinic cholinergic receptors (IC50 ∽ 0.03 μM for inhibition of catecholamine secretion in nicotine-stimulated PC-12 cells).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 137592-45-1
Molecular Formula C27H26N4O2
Molecular Weight (g/mol) 438.531
InChI Key LBFDERUQORUFIN-UHFFFAOYSA-N
Synonym bisindolylmaleimide ii, 2-1-2-1-methylpyrrolidino ethyl-1h-indol-3-yl-3-1h-indol-3-yl maleimide, bisindolyl deriv. 22, bim-2, bisindolylmaleimide ii mixture of 2 isomers, 3-1h-indol-3-yl-4-1-2-1-methylpyrrolidin-2-yl ethyl indol-3-yl pyrrole-2,5-dione, 3-1h-indol-3-yl-4-1-2-1-methyl-2-pyrrolidinyl ethyl-1h-indol-3-yl-1h-pyrrole-2,5-dione, 3-1h-indol-3-yl-4-1-2-1-methylpyrrolidin-2-yl ethyl-1h-indol-3-yl-2,5-dihydro-1h-pyrrole-2,5-dione, 3-1h-indol-3-yl-4-1-2-1-methylpyrrolidin-2-yl ethyl indol-3-yl-1h-pyrrole-2,5-dione
PubChem CID 2397
IUPAC Name 3-(1H-indol-3-yl)-4-[1-[2-(1-methylpyrrolidin-2-yl)ethyl]indol-3-yl]pyrrole-2,5-dione
SMILES CN1CCCC1CCN2C=C(C3=CC=CC=C32)C4=C(C(=O)NC4=O)C5=CNC6=CC=CC=C65

Specifications

Quantity 10 mg
Formula Weight 438.52
Percent Purity >97%
Chemical Name or Material Bisindolylmaleimide II
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.