Learn More
Enzo Life Sciences Bisindolylmaleimide III (5mg)
SDP
Catalog No. 502008384
Encompass_Preferred
Potent and selective inhibitor of protein kinase C (PKC). Formula: C23H20N4O2. MW: 384.4. Purity: ≥98%. Solubility: Soluble in methanol or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8384 Supplier Enzo Life Sciences Supplier No. ALX270051M005
Potent and selective inhibitor of protein kinase C (PKC). Formula: C23H20N4O2. MW: 384.4. Purity: ≥98%. Solubility: Soluble in methanol or DMSO. Long Term Storage: -20°C.