Learn More
AdipoGen Bisindolylmaleimide III (5 mg)
SDP
Catalog No. 502053839
Encompass_Preferred
Chemical. CAS: 137592-43-9. Formula: C23H20N4O2. MW: 384.4. Potent and selective protein kinase C (PKC) inhibitor. Inhibits panel of protein kinases (e.g. GSK-3beta, CDK2).
Catalog No. 50-205-3839 Supplier AdipoGen Supplier No. AGCR10112M005
Chemical. CAS: 137592-43-9. Formula: C23H20N4O2. MW: 384.4. Potent and selective protein kinase C (PKC) inhibitor. Inhibits panel of protein kinases (e.g. GSK-3beta, CDK2).