Learn More
Enzo Life Sciences Bisindolylmaleimide IV (1mg). CAS: 119139-23-0
SDP
Catalog No. 502008385
Encompass_Preferred
PKC inhibitor. Alternative name: Ro 31-6233, Arcyriarubin A. Formula: C20H13N3O2. MW: 327.3. Purity: ≥95% (HPLC), single spot (TLC). Appearance: Red solid. Solubility: Soluble in DMSO or methanol. Long Term Storage: -20°C. Handling: Protect from light.
Catalog No. 50-200-8385 Supplier Enzo Life Sciences Supplier No. ALX270052M001
PKC inhibitor. Alternative name: Ro 31-6233, Arcyriarubin A. Formula: C20H13N3O2. MW: 327.3. Purity: ≥95% (HPLC), single spot (TLC). Appearance: Red solid. Solubility: Soluble in DMSO or methanol. Long Term Storage: -20°C. Handling: Protect from light.