Learn More
AdipoGen Bisindolylmaleimide IV (1 mg)
SDP
Supplier:  AdipoGen AGCR10152M001
Encompass_Preferred
Chemical. CAS: 119139-23-0. Formula: C20H13N3O2. MW: 327.3. Cell permeable protein kinase C (PKC) inhibitor. Inhibits also protein kinase A (PKA).
Catalog No. 50-205-3854