Learn More
Cayman Chemical BIsIndolylmaleImIde IV 25mg
SDP
Catalog No. 502952923
Encompass_Preferred
A cell permeable inhibitor of PKC with IC50 values reported to range from 0.10 to 0.55 µM; designed to be more discriminative than its parent compound, the non-selective PKC inhibitor, staurosporine (Item No. 81590); also inhibits protein kinase A with IC50 values ranging from 2 to 11.8 µM
Catalog No. 50-295-2923 Supplier Cayman Chemical Supplier No. 1329925 MG
A cell permeable inhibitor of PKC with IC50 values reported to range from 0.10 to 0.55 µM; designed to be more discriminative than its parent compound, the non-selective PKC inhibitor, staurosporine (Item No. 81590); also inhibits protein kinase A with IC50 values ranging from 2 to 11.8 µM