Learn More
Enzo Life Sciences Bisindolylmaleimide IV (5mg). CAS: 119139-23-0
SDP
Supplier:  Enzo Life Sciences ALX270052M005
Encompass_Preferred
Alternative name: Ro 31-6233, Arcyriarubin A. Formula: C20H13N3O2. MW: 327.3. Purity: ≥95% (HPLC), single spot (TLC). Appearance: Red solid. Solubility: Soluble in DMSO or methanol. Long Term Storage: -20°C. Handling: Protect from light.
Catalog No. 50-200-8386