Learn More
Medchemexpress LLC Bisindolylmaleimide IV (Arcyriarubin A) | 119139-23-0 | MFCD00236432 | ≥98.0% | 327.34 g/mol | C20H13N3O2 | 5 MG
SDP
Catalog No. 5000243962
Encompass_Preferred
Bisindolylmaleimide IV (Arcyriarubin A) is a small-molecule inhibitor of protein kinase C (PKC) used in biochemical and cell-based research. It exhibits potent PKC inhibition (IC50 ~0.1-0.55 μM) and has reported antiviral activity against human cytomegalovirus (cell-culture IC50 ≈0.2 μM). The compound is supplied as a solid or as a DMSO solution for convenient in vitro dosing and is intended for research use only.
  • Potent protein kinase C inhibitor with low-micromolar IC50 values.
  • Reported inhibitor of human cytomegalovirus replication in cell culture.
  • Available as a solid and as a 10 mM solution in DMSO for convenient dosing.
  • Suitable for biochemical kinase assays and cell-based signaling studies.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-43962 Supplier Medchemexpress LLC Supplier No. HY1082545MG
Bisindolylmaleimide IV (Arcyriarubin A) is a small-molecule inhibitor of protein kinase C (PKC) used in biochemical and cell-based research. It exhibits potent PKC inhibition (IC50 ~0.1-0.55 μM) and has reported antiviral activity against human cytomegalovirus (cell-culture IC50 ≈0.2 μM). The compound is supplied as a solid or as a DMSO solution for convenient in vitro dosing and is intended for research use only.
  • Potent protein kinase C inhibitor with low-micromolar IC50 values.
  • Reported inhibitor of human cytomegalovirus replication in cell culture.
  • Available as a solid and as a 10 mM solution in DMSO for convenient dosing.
  • Suitable for biochemical kinase assays and cell-based signaling studies.
  • For research use only; not for human or veterinary use.