Learn More
Abcam Bisindolylmaleimide IV, PKC inhibitor, 1MG
Catalog No. 502705991
Encompass_Preferred
MW 327.3 Da, Purity >98%. Potent cell-permeable PKC inhibitor (IC₅₀ values are 0.1 - 0.55 μM). Inhibits PKA (IC₅₀ values are 2 - 11.8 μM). Noncompetitive nAChR antagonist. Shows less potent antinicotinic effects than Bisindolylmaleimide II (ab144206). Induces apoptosis and shows antiproliferative effects.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5991 Supplier Abcam Supplier No. AB1442081MG
MW 327.3 Da, Purity >98%. Potent cell-permeable PKC inhibitor (IC₅₀ values are 0.1 - 0.55 μM). Inhibits PKA (IC₅₀ values are 2 - 11.8 μM). Noncompetitive nAChR antagonist. Shows less potent antinicotinic effects than Bisindolylmaleimide II (ab144206). Induces apoptosis and shows antiproliferative effects.
The product is subject to the following: Abcam Restricted Use Statement