Learn More
Medchemexpress LLC Bisindolylmaleimide IX formic | 00-00-0 | 96.0% | 503.57 g/mol | C26H25N5O4S | 5 MG
SDP
Catalog No. 5000223440
Encompass_Preferred
Ro 31-8220 formic is the formic acid salt of bisindolylmaleimide IX, a potent protein kinase C (PKC) inhibitor provided for laboratory research. The compound is supplied as a small, high-purity solid with manufacturer storage and handling guidance available on the product datasheet.
  • Potent PKC inhibitor with low-nanomolar activity.
  • Formic acid salt for improved handling and solubility options.
  • High purity (≈96.0%).
  • Orange to red solid appearance.
  • Supplied as a 5 mg vial for research use.
  • Recommended storage: sealed, away from moisture, at -20 °C; in solvent, store at -80 °C (up to 6 months) or -20 °C (up to 1 month).
Catalog No. 50-002-23440 Supplier Medchemexpress LLC Supplier No. HY13866B5MG
Ro 31-8220 formic is the formic acid salt of bisindolylmaleimide IX, a potent protein kinase C (PKC) inhibitor provided for laboratory research. The compound is supplied as a small, high-purity solid with manufacturer storage and handling guidance available on the product datasheet.
  • Potent PKC inhibitor with low-nanomolar activity.
  • Formic acid salt for improved handling and solubility options.
  • High purity (≈96.0%).
  • Orange to red solid appearance.
  • Supplied as a 5 mg vial for research use.
  • Recommended storage: sealed, away from moisture, at -20 °C; in solvent, store at -80 °C (up to 6 months) or -20 °C (up to 1 month).