Learn More
Medchemexpress LLC Bisindolylmaleimide IX formic | 00-00-0 | 96.0% | C26H25N5O4S | 10MG
SDP
Catalog No. 5000260584
Encompass_Preferred
Ro 31-8220 formic is a small-molecule research reagent that acts as a potent, cell-permeable inhibitor of protein kinase C (PKC) and several other kinases. It is used in biochemical and cellular studies to probe PKC-dependent signaling, kinase selectivity, and downstream pathway effects such as JNK activation and c-Jun induction.
  • Potent PKC inhibition with low-nanomolar IC50s.
  • Inhibits multiple kinases for broad pathway interrogation.
  • Cell-permeable for use in cellular assays.
  • High purity solid with characteristic orange to red color.
  • Stable when stored sealed at -20°C; in solvent can be stored at -80°C for extended periods.
  • Available in small milligram pack sizes suitable for research use.
Catalog No. 50-002-60584 Supplier Medchemexpress LLC Supplier No. HY13866B10MG
Ro 31-8220 formic is a small-molecule research reagent that acts as a potent, cell-permeable inhibitor of protein kinase C (PKC) and several other kinases. It is used in biochemical and cellular studies to probe PKC-dependent signaling, kinase selectivity, and downstream pathway effects such as JNK activation and c-Jun induction.
  • Potent PKC inhibition with low-nanomolar IC50s.
  • Inhibits multiple kinases for broad pathway interrogation.
  • Cell-permeable for use in cellular assays.
  • High purity solid with characteristic orange to red color.
  • Stable when stored sealed at -20°C; in solvent can be stored at -80°C for extended periods.
  • Available in small milligram pack sizes suitable for research use.