Learn More
Medchemexpress LLC Bisindolylmaleimide V | 113963-68-1 | 97.75% | 341.36 | 100 MG
SDP
Catalog No. 5000438240
Encompass_Preferred
Bisindolylmaleimide V is a cell-permeable negative control for protein kinase C inhibition studies, exhibiting an IC50 value exceeding 100 μM. It effectively blocks the activation of mitogen-stimulated protein kinase p70s6k/p85s6k (S6K) in vivo, with an IC50 of 8 μM.
  • Cell-permeable negative control for protein kinase C inhibition studies
  • Exhibits an IC50 value exceeding 100 μM for protein kinase C inhibition
  • Effectively blocks the activation of mitogen-stimulated protein kinase p70s6k/p85s6k (S6K) in vivo
  • Has an IC50 of 8 μM for S6K activation blocking
Catalog No. 50-004-38240 Supplier Medchemexpress LLC Supplier No. HY112400100MG
Bisindolylmaleimide V is a cell-permeable negative control for protein kinase C inhibition studies, exhibiting an IC50 value exceeding 100 μM. It effectively blocks the activation of mitogen-stimulated protein kinase p70s6k/p85s6k (S6K) in vivo, with an IC50 of 8 μM.
  • Cell-permeable negative control for protein kinase C inhibition studies
  • Exhibits an IC50 value exceeding 100 μM for protein kinase C inhibition
  • Effectively blocks the activation of mitogen-stimulated protein kinase p70s6k/p85s6k (S6K) in vivo
  • Has an IC50 of 8 μM for S6K activation blocking