Learn More
Medchemexpress LLC Bisindolylmaleimide V | 113963-68-1 | 97.75% | 341.36 | 500 MG
SDP
Catalog No. 5000438482
Encompass_Preferred
Bisindolylmaleimide V is a cell-permeable negative control for protein kinase C inhibition studies, with an IC50 value exceeding 100 μM. It also blocks the activation of mitogen-stimulated protein kinase p70s6k/p85s6k (S6K) in vivo, with an IC50 of 8 μM. This product is intended for research use only and is not sold to patients.
  • Cell-permeable negative control for protein kinase C inhibition studies.
  • Blocks activation of mitogen-stimulated protein kinase p70s6k/p85s6k (S6K) in vivo.
Catalog No. 50-004-38482 Supplier Medchemexpress LLC Supplier No. HY112400500MG
Bisindolylmaleimide V is a cell-permeable negative control for protein kinase C inhibition studies, with an IC50 value exceeding 100 μM. It also blocks the activation of mitogen-stimulated protein kinase p70s6k/p85s6k (S6K) in vivo, with an IC50 of 8 μM. This product is intended for research use only and is not sold to patients.
  • Cell-permeable negative control for protein kinase C inhibition studies.
  • Blocks activation of mitogen-stimulated protein kinase p70s6k/p85s6k (S6K) in vivo.