Learn More
Medchemexpress LLC Bisindolylmaleimide V | 113963-68-1 | MFCD00236433 | 97.8% | 341.36 | C21H15N3O2 | 5 MG
SDP
Catalog No. 5000264869
Encompass_Preferred
Bisindolylmaleimide V is a synthetic small-molecule research reagent used as a cell-permeable negative control in kinase signaling studies, particularly for protein kinase C and S6 kinase pathways. It is supplied at research scales and is characterized by a reported purity of 97.75%.
  • Cell-permeable negative control for protein kinase C inhibition studies (IC50 >100 μM).
  • Reported to block activation of p70/p85 S6 kinase in vivo (IC50 8 μM).
  • Molecular formula C21H15N3O2 and molecular weight 341.36.
  • Reported purity 97.8%.
  • Supplied in research-scale quantities, including 5 mg.
Catalog No. 50-002-64869 Supplier Medchemexpress LLC Supplier No. HY1124005MG
Bisindolylmaleimide V is a synthetic small-molecule research reagent used as a cell-permeable negative control in kinase signaling studies, particularly for protein kinase C and S6 kinase pathways. It is supplied at research scales and is characterized by a reported purity of 97.75%.
  • Cell-permeable negative control for protein kinase C inhibition studies (IC50 >100 μM).
  • Reported to block activation of p70/p85 S6 kinase in vivo (IC50 8 μM).
  • Molecular formula C21H15N3O2 and molecular weight 341.36.
  • Reported purity 97.8%.
  • Supplied in research-scale quantities, including 5 mg.