Learn More
AdipoGen Bisindolylmaleimide V (1 mg)
SDP
Supplier:  AdipoGen AGCR10023M001
Encompass_Preferred
Chemical. CAS: 113963-68-1. Formula: C21H15N3O2. MW: 341.4. Negative control for protein kinase C (PKC) inhibitors. S6K inhibitor. Necrosis inhibitor.
Catalog No. 50-205-3772