Learn More
Enzo Life Sciences Bisindolylmaleimide V (5mg). CAS: 113963-68-1
SDP
Catalog No. 502008388
Encompass_Preferred
Negative control for the protein kinase C (PKC)-inhibitory activity. Formula: C21H15N3O2. MW: 341.4. Purity: ≥98%. Solubility: Soluble in DMSO or methanol; very slightly soluble in 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-200-8388 Supplier Enzo Life Sciences Supplier No. ALX270053M005
Add to Cart
Add to Cart
Negative control for the protein kinase C (PKC)-inhibitory activity. Formula: C21H15N3O2. MW: 341.4. Purity: ≥98%. Solubility: Soluble in DMSO or methanol; very slightly soluble in 100% ethanol. Long Term Storage: -20°C.