Learn More
TARGETMOL CHEMICALS INC Bisindolylmaleimide X hydrochl
SDP
Catalog No. 502880580
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Bisindolylmaleimide X hydrochloride (BIM-X hydrochloride) is a potent and selective PKC inhibitor and a CDK2 antagonist with an IC50 of 200 nM. Purity 98.5%
Catalog No. 50-288-0580 Supplier TARGETMOL CHEMICALS INC Supplier No. T105505MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Bisindolylmaleimide X hydrochloride (BIM-X hydrochloride) is a potent and selective PKC inhibitor and a CDK2 antagonist with an IC50 of 200 nM. Purity 98.5%