Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bisindolylmaleimide XI hydrochloride | 145333-02-4 | MFCD01319127 | 98.0% | 489.01 g/mol | C28H29ClN4O2 | 1 MG
SDP

Catalog No. 5000227670
Encompass_Preferred

Bisindolylmaleimide XI hydrochloride is a bisindolylmaleimide-class small molecule and a potent inhibitor of protein kinase C (PKC) isoforms. It is supplied as the hydrochloride salt for research use and is used in studies of PKC-dependent signaling and cellular pathways.

  • Potent PKC inhibitor for biochemical and cellular studies.
  • Hydrochloride salt form improves solubility for assay preparation.
  • Available in small research pack sizes (1 mg, 5 mg, 10 mg).
  • Molecular weight 489.01 g/mol; formula C28H29ClN4O2.
  • High purity (98.0%) suitable for research applications.

Catalog No. 50-002-27670 Supplier Medchemexpress LLC Supplier No. HY117610A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bisindolylmaleimide XI hydrochloride is a bisindolylmaleimide-class small molecule and a potent inhibitor of protein kinase C (PKC) isoforms. It is supplied as the hydrochloride salt for research use and is used in studies of PKC-dependent signaling and cellular pathways.

  • Potent PKC inhibitor for biochemical and cellular studies.
  • Hydrochloride salt form improves solubility for assay preparation.
  • Available in small research pack sizes (1 mg, 5 mg, 10 mg).
  • Molecular weight 489.01 g/mol; formula C28H29ClN4O2.
  • High purity (98.0%) suitable for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.