Learn More
Medchemexpress LLC Bismaleimide | 13676-54-5 | 25 G
SDP
Catalog No. 5000233271
Encompass_Preferred
Bismaleimide (CAS 13676-54-5) is an aromatic bismaleimide reagent (C21H14N2O4, MW 358.35 g/mol) used as a crosslinking and heat-resistant bridging unit in polymer and materials research. Supplied as a 25 g package for laboratory use.
  • Aromatic bismaleimide structure suitable for crosslinking.
  • Provides thermal stability in polymer and aerogel formulations.
  • Solid reagent supplied in small laboratory pack sizes.
  • Applicable to materials chemistry and biochemical assay workflows.
  • Molecular weight 358.35 g/mol; chemical formula C21H14N2O4.
Catalog No. 50-002-33271 Supplier Medchemexpress LLC Supplier No. HY11952625G
Bismaleimide (CAS 13676-54-5) is an aromatic bismaleimide reagent (C21H14N2O4, MW 358.35 g/mol) used as a crosslinking and heat-resistant bridging unit in polymer and materials research. Supplied as a 25 g package for laboratory use.
  • Aromatic bismaleimide structure suitable for crosslinking.
  • Provides thermal stability in polymer and aerogel formulations.
  • Solid reagent supplied in small laboratory pack sizes.
  • Applicable to materials chemistry and biochemical assay workflows.
  • Molecular weight 358.35 g/mol; chemical formula C21H14N2O4.