Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bismaleimide | 13676-54-5 | 96.6% | 100 G
SDP

Catalog No. 5000234234
Encompass_Preferred

Bismaleimide (CAS 13676-54-5) is a light yellow solid bifunctional maleimide used in organic synthesis, crosslinking, and polymer modification. It is supplied with analytical purity suitable for research applications and is soluble in DMSO.

  • CAS number: 13676-54-5.
  • Purity: 96.6%.
  • Molecular weight: 358.35 g/mol.
  • Chemical formula: C21H14N2O4.
  • Solubility: DMSO 100 mg/mL; may require sonication.
  • Appearance: light yellow solid.
  • Recommended storage: store at room temperature; in solvent, store at -20°C to -80°C.

Catalog No. 50-002-34234 Supplier Medchemexpress LLC Supplier No. HY119526100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bismaleimide (CAS 13676-54-5) is a light yellow solid bifunctional maleimide used in organic synthesis, crosslinking, and polymer modification. It is supplied with analytical purity suitable for research applications and is soluble in DMSO.

  • CAS number: 13676-54-5.
  • Purity: 96.6%.
  • Molecular weight: 358.35 g/mol.
  • Chemical formula: C21H14N2O4.
  • Solubility: DMSO 100 mg/mL; may require sonication.
  • Appearance: light yellow solid.
  • Recommended storage: store at room temperature; in solvent, store at -20°C to -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.