Learn More
Medchemexpress LLC HY-118567 10mg Medchemexpress, Bis(maltolato)oxovanadium(IV) CAS:38213-69-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11856710MG
Encompass_Preferred
Medchemexpress, HY-118567 10mg Bis(maltolato)oxovanadium(IV) CAS:38213-69-3 Bis(maltolato)oxovanadium(IV) (BMOV) is a potent, reversible, competitive and orally active pan-PTP (protein tyrosine phosphatases) inhibitor. Bis(maltolato)oxovanadium(IV) inhibits HCPTPA, PTP1B, HPTPβ and SHP2 with IC50s of 126 nM, 109 nM, 26 nM and 201 nM, respectively. Bis(maltolato)oxovanadium(IV) is a potent insulin sensitizer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0916