Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Bismarck Brown Y 25 GR
SDP

Catalog No. 50980200
Encompass_Preferred

(Basic Brown 1; Basic Brown G, GX, GXP; Vesuvine; Leather Brown; Manchester Brown; Phenylene Brown)
CAS #10114-58-6 C₁₈H₁₈N·2HCl F.W. 419.31
Solubility: 1.5% Water; 3.0% Alcohol; 3.0% Cellosolve; 7.9% Glycol; 0.05% Xylene

PAP for staining smears. Nuclei & granules. Mucin & calciform cells of intestine, cartilage & embryo.

Catalog No. 50-980-200 Supplier Electron Microscopy Sciences Supplier No. 11310
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Basic Brown 1; Basic Brown G, GX, GXP; Vesuvine; Leather Brown; Manchester Brown; Phenylene Brown)
CAS #10114-58-6 C₁₈H₁₈N·2HCl F.W. 419.31
Solubility: 1.5% Water; 3.0% Alcohol; 3.0% Cellosolve; 7.9% Glycol; 0.05% Xylene

PAP for staining smears. Nuclei & granules. Mucin & calciform cells of intestine, cartilage & embryo.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.