Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismarck Brown Y MP Biomedicals

Catalog No. ICN15047380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15047380 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15047380 Supplier MP Biomedicals, Inc Supplier No. 0215047380
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Plasma stain used for staining mucine, cartilage, and goblet cells.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10114-58-6
Molecular Formula C18H20Cl2N8
Molecular Weight (g/mol) 419.314
InChI Key MCZVRBLCRZWFJH-UHFFFAOYSA-N
Synonym 4,4-[1,3-PhenyleneBis(Azo)Bis]-1,3-Benzediamine Dihydrochloride, Basic Brown 1
PubChem CID 82360
ChEBI CHEBI:53615
IUPAC Name 4-[[3-[(2,4-diaminophenyl)diazenyl]phenyl]diazenyl]benzene-1,3-diamine;dihydrochloride
SMILES C1=CC(=CC(=C1)N=NC2=C(C=C(C=C2)N)N)N=NC3=C(C=C(C=C3)N)N.Cl.Cl

Specifications

Assay Percent Range ∽ 50%
Color Orange
Quantity 100 g
Formula Weight 419.3
Grade Not Available
Chemical Name or Material Bismarck Brown Y, Plasma stain for mucin, cartilage and goblet cells
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.