Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth aluminum oxide hydrate, 95.5%, Thermo Scientific™

Catalog No. AA4039330
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4039330 250 g
AA4039318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4039330 Supplier Thermo Scientific Chemicals Supplier No. 4039330
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00798526
Synonym Aluminum bismuth oxide hydrate; Bismuth aluminate hydrate

Specifications

Physical Form Powder
Quantity 250 g
Solubility Information Insoluble in water; Soluble in nitric acid; Typical tap density 0;69g/cm3
Formula Weight 771.84 (Anhydrous)
Percent Purity 95.5%
Chemical Name or Material Bismuth aluminum oxide hydrate
EINECSNumber 235-552-3
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.