Learn More
Electron Microscopy Sciences Bismuth Carbonate, Basic, Purified 25 GR
SDP
Catalog No. 50980203
Encompass_Preferred
(Bismuth Subcarbonate)
CAS #5892-10-4 Approx. (BiO)₂CO₃ F.W. 509.96
Used for staining synapses. Pfenninger, K., Sandri, C., Albert, K. and Eugster, C.H. (1969). Contribution of the problem of structural organization of the presynaptic area. Brain Res., 12:10
Catalog No. 50-980-203 Supplier Electron Microscopy Sciences Supplier No. 11420
(Bismuth Subcarbonate)
CAS #5892-10-4 Approx. (BiO)₂CO₃ F.W. 509.96
Used for staining synapses. Pfenninger, K., Sandri, C., Albert, K. and Eugster, C.H. (1969). Contribution of the problem of structural organization of the presynaptic area. Brain Res., 12:10