Learn More
Strem, An Ascensus Company CAS# 67874-71-9. 1kg. Bismuth (III) 2-ethylhexanoate mineral spirits (approx. 28% Bi). MFCD00043803
SDP
Catalog No. 5090113545
Encompass
CAS# 67874-71-9. 1kg. Bismuth (III) 2-ethylhexanoate mineral spirits (approx. 28% Bi). MFCD00043803. Molecular Weight: 638.61. Molecular Formula: Bi[OOCCH(C2H5)C4H9]3. Color/form: brown liq. Strem# 83-2400. http://www.strem.com/catalog/v/83-2400/
Catalog No. 50-901-13545 Supplier Strem, An Ascensus Company Supplier No. 8324001000G
CAS# 67874-71-9. 1kg. Bismuth (III) 2-ethylhexanoate mineral spirits (approx. 28% Bi). MFCD00043803. Molecular Weight: 638.61. Molecular Formula: Bi[OOCCH(C2H5)C4H9]3. Color/form: brown liq. Strem# 83-2400. http://www.strem.com/catalog/v/83-2400/