Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bismuth(III) Acetate

Catalog No. 40158750G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-158-750G 50 g
40-158-710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 40-158-750G Supplier Sigma Aldrich Supplier No. 40158750G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Bismuth(III) acetate is employed as a starting material in the synthesis of bismuth(III) sulfide (Bi2S3) which can be used in solution-processable bulk heterojunction solar cells.It is used in the preparation of gold-bismuth sulfide (Au-Bi2S3) heteronanostructures and bismuth titanate nanorods as photocatalysts.It is also used in the synthesis of triarylbismuth compounds.

Specifications

CAS 22306-37-2
Molecular Formula C6H9BiO6
Linear Formula (CH3CO2)3Bi
MDL Number MFCD00015629
Quantity 50 g
Synonym Bismuth triacetate; Acetic acid bismuth(III) salt
Molecular Weight (g/mol) 386.11 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.