Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bismuth(III) Gallate Basic Hydrate

Catalog No. 15102100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-102-100G 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-102-100G Supplier Sigma Aldrich Supplier No. 15102100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Investigations of: Pharmacological activity, specifically NO and PGE2 production by macrophagesTreatment of Helicobacter pylori infections

Specifications

CAS 201467-81-4
Density 0.750 to 0.850 g/mL at 20°C
Molecular Formula C10H19N3O6
Linear Formula C10H19N3O6
MDL Number MFCD20134123
Quantity 500 mg
Synonym 2,7-Dihydroxy-1,3,2-benzodioxabismole-5-carboxylic acid; Bismuth subgallate; Dermatol
Molecular Weight (g/mol) 277.27 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.