Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth(III) iodide, ultra dry, 99.998% (metals basis)

Catalog No. AA3568106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3568106 5 g
AA3568114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3568106 Supplier Thermo Scientific Chemicals Supplier No. 03568106
Add to Cart

CAS: 7787-64-6 | BiI3 | 589.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-64-6
Molecular Formula BiI3
Molecular Weight (g/mol) 589.69
MDL Number MFCD00010894
InChI Key KOECRLKKXSXCPB-UHFFFAOYSA-K
Synonym bismuth iodide, bismuth iii iodide, bismuth triiodide, bismuthine, triiodo, triiodobismuthine, triiodobismuth, bismuth iodide,, ksc179g5d
PubChem CID 111042
IUPAC Name bismuth(3+) triiodide
SMILES [I-].[I-].[I-].[Bi+3]

Specifications

Boiling Point ∼500°C
Melting Point 408°C
Physical Form Granules
Quantity 5 g
UN Number UN3260
Merck Index 14,1269
Formula Weight 589.69
Percent Purity 99.998%
Odor Acidic
Packaging Ampule
Sensitivity Hygroscopic
Density 5.778 g/mL
Assay (metals basis)
Chemical Name or Material Bismuth(III) iodide, Ultra dry
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 232-127-4
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.