Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bismuth(III) oxide

Catalog No. 223891100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-389-1100G 100 g
22-389-1500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-389-1100G Supplier Sigma Aldrich Supplier No. 223891100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Bismuth (III) oxide is a yellow, monoclinic crystalline powder. It is insoluble in water and hydroxide solutions but dissolves in acids to form bismuth (III) salts. It can be prepared by heating bismuth in air or by heating hydroxides, carbonates or nitrates of bismuth.General applications of bismuth (III) oxide are:production of optical glasses with high indices of refraction, precursor in the production of other bismuth compounds. Bismuth (III) oxide is suitable for dental materials to render them opaque to X-rays.It is used for the synthesis of pure BiFeO3 perovskite nanoparticles, lead free piezoelectric 0.96(Bi0.5Na0.5)TiO3 - 0.04 BaTiO3.ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 1304-76-3
Quantity 100 g
Molecular Formula Bi2O3
Linear Formula Bi2O3
MDL Number MFCD00003462
Synonym Dibismuth trioxide
Molecular Weight (g/mol) 465.96
Percent Purity 99.9% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.