Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth(III) sulfate, 99%, pure, Thermo Scientific Chemicals

Catalog No. AC453850250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453850250 25 g
AC453851000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC453850250 Supplier Thermo Scientific Chemicals Supplier No. 453850250
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-68-0
Molecular Formula Bi2O12S3
Molecular Weight (g/mol) 706.13
MDL Number MFCD00036140
InChI Key BEQZMQXCOWIHRY-UHFFFAOYSA-H
Synonym bismuth sulfate, dibismuth trisulfate, bismuth trisulfate, sulfuric acid, bismuth 3+ salt 3:2, bismuth sulphate, bismuth sesquisulfate, bismuth sulfate mi, dibismuth tris sulphate, bismuth iii sulfate, 2bi.3so4
PubChem CID 198144
IUPAC Name dibismuth;trisulfate
SMILES [Bi+3].[Bi+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Melting Point 405.0°C
Quantity 25 g
Assay Percent Range 98.5% min
Linear Formula Bi2(SO4)3
Merck Index 15,1286
Formula Weight 706.13
Percent Purity 99%
Grade Pure
Chemical Name or Material Bismuth(III) sulfate
EINECSNumber 232-129-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.